Precrimped, Crimp-Top Vials, Large Opening, 11 mm, Thermo Scientific
Supplier: Thermo Scientific
Certificates
About this item
Glass vials are precrimped with an aluminum seal and PTFE/red rubber septum.
- Aluminum seal is crimped onto the vial following a purge with dry nitrogen
- Sample is added via injection through the septum
Save time during sample preparation and reduce the risk of contamination.
Packaging: Packaged in convenient vial trays with clear covers.
Customers Who Bought This Also Bought