700 System Luer-Lock Tip Caps, Techcon
700 System Luer-Lock Tip Caps
Catalog # 14219-636
Supplier: Techcon Systems
Supplier #: TE7015LBPK
Jump to:
Product Details & Documents
Tip caps allow products to be interchanged without compromising vital UV block protection
Made of polyethylene. Use with 700 System barrels (see 14219-652 series) to seal barrels between usage or to seal any valve. Color: black.
Specifications
- Description:700 System Luer-Lock Tip Caps