Orders LinkContactUsLinkComponent
VWR® Spectrosil Fluorometer Cells
VWR® Spectrosil Fluorometer Cells
VWR® Spectrosil Fluorometer Cells
Supplier:  VWR International
Certificates
VWR® Spectrosil Fluorometer Cells
VWR® Spectrosil Fluorometer Cells
4 Options Available Below
Product Details & Documents
Standard rectangular cells manufactured from Spectrosil quartz for accurate readings over the entire UV range to near infrared range.
  • Far UV quartz
  • Wavelength range of 170–2700 nm
  • Fully heat-fused construction
Due to the fully heat-fused construction (no intermediate materials or adhesives) and careful annealing to remove any strain, there is no optical or physical distortion of the four optically polished windows
Lids and stoppers are made of polytetrafluoroethylene (PTFE). Cell 414004-057 features a closed-top polymer screw cap with a silicone rubber gasket. Cell 414004-058 features an open-top polymer screw cap with a silicone rubber septum.
Cells exhibit a transmission tolerance within 1.5% at 200nm.
Ordering information: For Spectrosil cells for use in spectrophotometers, see 414004-060 series.
Product Family Options
Product Information
  • Internal dimensions
    Sample volume
    Pathlength
    Cover type
    Ext. dimensions
    Availability
    Price
  • W×L 10×10 mm
    3.5 ml
    10 mm
    Stopper
    12.5W x 12.5L x 45H mm
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $540.34
    $540.34
    New Customer Promotional Price
    Log in for availability
    Case of 8
    Case of 8 - $4,322.69
    $4,322.69
    New Customer Promotional Price
    Pack of 1
       $540.34
    Case of 8
       $4,322.69
  • W×L 10×10 mm
    3.5 ml
    10 mm
    Flat lid
    12.5W x 12.5L x 45H mm
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $1,864.87
    $1,864.87
    New Customer Promotional Price
    Log in for availability
    Case of 16
    Case of 16 - $14,918.98
    $14,918.98
    New Customer Promotional Price
    Pack of 2
       $1,864.87
    Case of 16
       $14,918.98
  • W×L 10×10 mm
    3.5 ml
    10 mm
    Screw cap
    12.5W x 12.5L x 52H mm
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $558.19
    $558.19
    New Customer Promotional Price
    Log in for availability
    Case of 8
    Case of 8 - $4,465.51
    $4,465.51
    New Customer Promotional Price
    Pack of 1
       $558.19
    Case of 8
       $4,465.51
  • W×L 10×10 mm
    3.5 ml
    10 mm
    Septum screw cap
    12.5W x 12.5L x 52H mm
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $777.82
    $777.82
    New Customer Promotional Price
    Log in for availability
    Case of 8
    Case of 8 - $6,222.59
    $6,222.59
    New Customer Promotional Price
    Pack of 1
       $777.82
    Case of 8
       $6,222.59