ContactUsLinkComponent
Double Lens Magnifier, Bausch & Lomb®
Double Lens Magnifier, Bausch & Lomb®
Catalog # 36933-020
Supplier:  Avantor
undefined
Double Lens Magnifier, Bausch & Lomb®
Catalog # 36933-020
Supplier:  Avantor
Specifications
  • Ø:
    16 mm (15/8")
  • Magnification:
    3.5×
  • Cat. no.:
    36933-020
  • Supplier No.:
    813476
Specifications
About this item
Offers an unusually wide and flat field for 3.5X magnification
Two plano-convex lenses are used. Provides the corrected wide flat field.