VACUETTE® Blood Collection Tube Caps, Greiner Bio-One
Supplier: Greiner Bio-One
Certificates
About this item
Snap caps are suitable for recapping all 13mm VACUETTE® tubes.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.