ContactUsLinkComponent
VWR® Microcentrifuge Tube Rack, PP
VWR® Microcentrifuge Tube Rack, PP
Catalog # 20901-675
Supplier:  VWR International
undefined
VWR® Microcentrifuge Tube Rack, PP
Catalog # 20901-675
Supplier:  VWR International
Specifications
  • Dimensions:
    75 x 210 x 35 mm (2.9 x 8.3 x 1.4")
  • Cat. no.:
    20901-675
Specifications
About this item
Each polypropylene microcentrifuge tube rack stores 20 standard 1.5–2.0 mL microcentrifuge tubes in clearly numbered cavities.
  • Numbered cavities for quick identification
  • One-handed transportation
  • Autoclavable
  • Polypropylene construction
Easily transportable with one hand. Made of autoclavable polypropylene. Non-skid rubber feet.