DuPont™ ProShield® 60 Sleeves
Supplier: DuPont
Certificates
About this item
ProShield® 60 garments are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.
- Elastic at both ends
- Serged seams
- One size fits most
ProShield® 60 garments are made with a microporous film that is laminated to a spunbonded polypropylene substrate. Applications include automotive refinishing, waste cleanup and remediation.
These sleeves feature elastic at both biceps, measuring 83/4", and wrist, measuring 61/4".
Certifications: Thickness meets ASTM D751.
Specifications
- Catalog No:
- CA13452-326
- Garment application:
- Laboratory