Order Entry
Canada
ContactUsLinkComponent
Corning® Screw Cap with Plug Seal, Polypropylene, Corning
Corning® Screw Cap with Plug Seal, Polypropylene, Corning
Catalog # 89089-662
Supplier:  Corning
Product available on GSA Advantage®
undefined
Corning® Screw Cap with Plug Seal, Polypropylene, Corning
Catalog # 89089-662
Supplier:  Corning
Supplier Number:  1397-80LTC
Product available on GSA Advantage®

Specifications

  • Cap Size:
    GLS80
  • Environmentally Preferable:
  • Cat. no.:
    89089-662
  • Supplier No.:
    1397-80LTC

Specifications

About this item

Linerless orange cap has GLS80 thread and is autoclavable at 121°C (250°F).