ContactUsLinkComponent
SP Bel-Art Electrophoresis Gel Plate Rack, Bel-Art Products, a part of SP
SP Bel-Art Electrophoresis Gel Plate Rack, Bel-Art Products, a part of SP
Catalog # 77776-794
undefined
SP Bel-Art Electrophoresis Gel Plate Rack, Bel-Art Products, a part of SP
Catalog # 77776-794
Supplier Number:  F13596-0000
Specifications
  • Description:
    Electrophoresis Gel Plate Rack
  • Cat. no.:
    77776-794
  • Supplier No.:
    135960000
Specifications
About this item
This “A” shaped rack has ten places for holding glass plates and a weighted base for safety.
  • Polypropylene construction cushions glass edges
  • Secure transport without chipping or cracking
  • Small footprint
  • Weighted base to prevent tipping over
It features a round bottom and wide handles to provide a secure grip while transporting the rack. Smooth polypropylene material cushions glass edges to reduce chipping and cracking. A flat area on each end is designed for labels. The rack has a footprint of 20.3Wx38.1Dx22.7H cm (8x15x9") and weighs 1.7kg (33/4lbs). Available in white with steel fittings.