8 mm Crimp-Top Vials, Thermo Scientific
Supplier: Thermo Scientific
Certificates
About this item
Vials offer a custom fit for PerkinElmer® and Beckman Coulter® 504 autosamplers.
Made from clear 33 or amber 203 borosilicate glass. They are compatible with 8 mm aluminum crimp seals.
Customers Who Bought This Also Bought