Synthware Friedrichs Condensers, Kemtech America
Supplier: Kemtech America
Certificates
About this item
The top joint is an outer [ST] joint at a 15° angle.
- Heavy wall borosilicate construction
- Jacketed
- Ground glass joint
This Freidrichs condenser is extremely efficient and can ve used for reflux and distillation.
Frequently Bought Together