VWR® Basic SPP Frocks
About this item
Disposable frock made from spunbond polypropylene (SPP) fabric, offering lightweight and breathable protection for non-hazardous environments. Designed for comfort and contamination control, it features snap-front closure, non-linting material, and an economical single-use design.
Lightweight and breathable fabric reduces heat buildup during long shifts
Non-linting material helps maintain clean environments
Snap-front closure for easy donning and removal
Elastic cuffs for flexibility
Extended coverage compared to lab coats
Disposable, single-use design for contamination control
Economical solution for basic protection needs
The VWR® Basic Protection Frock is designed for environments where cleanliness and contamination control are essential, but exposure risks remain minimal. Made from spunbond polypropylene (SPP), this non-woven fabric offers a lightweight and breathable solution that ensures comfort during extended wear. Its non-linting properties help reduce particle contamination, making it suitable for clean areas, food processing, and general industrial applications.
The frock features a snap-front closure for quick and secure fastening, along with elastic cuffs to accommodate different glove types and ensure a secure fit. The disposable design supports hygiene compliance by minimizing cross-contamination and simplifying inventory management. While not intended for hazardous chemical or liquid protection, the frock provides an effective barrier against dust, dirt, and dry particulates, making it ideal for routine tasks in controlled environments. Combining cost-effectiveness, breathability, and basic contamination control, VWR® Basic Protection SPP Frocks deliver a practical solution for facilities seeking dependable single-use apparel for everyday operations.
: Frocks are packaged 30 per case.
5 Options Available Below
Product Details & Documents
Disposable frock made from spunbond polypropylene (SPP) fabric, offering lightweight and breathable protection for non-hazardous environments. Designed for comfort and contamination control, it features snap-front closure, non-linting material, and an economical single-use design.
Lightweight and breathable fabric reduces heat buildup during long shifts
Non-linting material helps maintain clean environments
Snap-front closure for easy donning and removal
Elastic cuffs for flexibility
Extended coverage compared to lab coats
Disposable, single-use design for contamination control
Economical solution for basic protection needs
The VWR® Basic Protection Frock is designed for environments where cleanliness and contamination control are essential, but exposure risks remain minimal. Made from spunbond polypropylene (SPP), this non-woven fabric offers a lightweight and breathable solution that ensures comfort during extended wear. Its non-linting properties help reduce particle contamination, making it suitable for clean areas, food processing, and general industrial applications.
The frock features a snap-front closure for quick and secure fastening, along with elastic cuffs to accommodate different glove types and ensure a secure fit. The disposable design supports hygiene compliance by minimizing cross-contamination and simplifying inventory management. While not intended for hazardous chemical or liquid protection, the frock provides an effective barrier against dust, dirt, and dry particulates, making it ideal for routine tasks in controlled environments. Combining cost-effectiveness, breathability, and basic contamination control, VWR® Basic Protection SPP Frocks deliver a practical solution for facilities seeking dependable single-use apparel for everyday operations.
: Frocks are packaged 30 per case.
Specifications for Product Family
Specifications
- Catalog No:
- 414004-384
- 414004-383
- 414004-381
- 414004-382
- 414004-400
- Fabric:
- Spunbound polypropylene
- Spunbound polypropylene
- Spunbound polypropylene
- Spunbound polypropylene
- Spunbound polypropylene
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Cleanroom class:
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- Pockets:
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Garment application:
- Cleanroom|Industrial|Laboratory
- Cleanroom|Industrial|Laboratory
- Cleanroom|Industrial|Laboratory
- Cleanroom|Industrial|Laboratory
- Cleanroom|Industrial|Laboratory
- Air permeability:
- 840.63 cu. ft./min/sq. ft.
- 840.63 cu. ft./min/sq. ft.
- 840.63 cu. ft./min/sq. ft.
- 840.63 cu. ft./min/sq. ft.
- 840.63 cu. ft./min/sq. ft.
- Bacterial filtration efficiency:
- 55%
- 55%
- 55%
- 55%
- 55%
- Particle shedding (Helmke Drum):
- Level II
- Level II
- Level II
- Level II
- Level II
- Water resistance:
- 21.7 G
- 21.7 G
- 21.7 G
- 21.7 G
- 21.7 G
- Water vapor transmission rate:
- 1257 g/m²/24 h
- 1257 g/m²/24 h
- 1257 g/m²/24 h
- 1257 g/m²/24 h
- 1257 g/m²/24 h
- Tensile strength:
- 25.9 lbs./in. (warp), 23.1 lbs./in. (filling)
- 25.9 lbs./in. (warp), 23.1 lbs./in. (filling)
- 25.9 lbs./in. (warp), 23.1 lbs./in. (filling)
- 25.9 lbs./in. (warp), 23.1 lbs./in. (filling)
- 25.9 lbs./in. (warp), 23.1 lbs./in. (filling)
- Tearing strength:
- 4.2 lbs./in. (lengthwise yarns), 4 lbs./in. (widthwise yarns)
- 4.2 lbs./in. (lengthwise yarns), 4 lbs./in. (widthwise yarns)
- 4.2 lbs./in. (lengthwise yarns), 4 lbs./in. (widthwise yarns)
- 4.2 lbs./in. (lengthwise yarns), 4 lbs./in. (widthwise yarns)
- 4.2 lbs./in. (lengthwise yarns), 4 lbs./in. (widthwise yarns)
- Weight:
- 48.82 g/m²
- 48.82 g/m²
- 48.82 g/m²
- 48.82 g/m²
- 48.82 g/m²
Specifications
Product Family Options
Product Information
- ColorClothing SizeLengthSleeveAvailabilityPrice
- WhiteM42½"34½"Log in for availabilityPallet of 720Pallet of 720 - $7,053.12$7,053.12New Customer Promotional PriceCase of 30$293.88Pallet of 720$7,053.12Specifications:
More
Specifications
Cat. No.414004-384FabricSpunbound polypropyleneClosure typePress studs / snapsCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroom|Industrial|LaboratoryAir permeability840.63 cu. ft./min/sq. ft.Bacterial filtration efficiency55%Particle shedding (Helmke Drum)Level IIWater resistance21.7 GWater vapor transmission rate1257 g/m²/24 hTensile strength25.9 lbs./in. (warp), 23.1 lbs./in. (filling)Tearing strength4.2 lbs./in. (lengthwise yarns), 4 lbs./in. (widthwise yarns)Weight48.82 g/m² - WhiteL43½"35½"Log in for availabilityPallet of 720Pallet of 720 - $7,525.63$7,525.63New Customer Promotional PriceCase of 30$313.57Pallet of 720$7,525.63Specifications:
More
Specifications
Cat. No.414004-383FabricSpunbound polypropyleneClosure typePress studs / snapsCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroom|Industrial|LaboratoryAir permeability840.63 cu. ft./min/sq. ft.Bacterial filtration efficiency55%Particle shedding (Helmke Drum)Level IIWater resistance21.7 GWater vapor transmission rate1257 g/m²/24 hTensile strength25.9 lbs./in. (warp), 23.1 lbs./in. (filling)Tearing strength4.2 lbs./in. (lengthwise yarns), 4 lbs./in. (widthwise yarns)Weight48.82 g/m² - WhiteXL46"36½"Log in for availabilityPallet of 720Pallet of 720 - $7,536.19$7,536.19New Customer Promotional PriceCase of 30$314.01Pallet of 720$7,536.19Specifications:
More
Specifications
Cat. No.414004-381FabricSpunbound polypropyleneClosure typePress studs / snapsCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroom|Industrial|LaboratoryAir permeability840.63 cu. ft./min/sq. ft.Bacterial filtration efficiency55%Particle shedding (Helmke Drum)Level IIWater resistance21.7 GWater vapor transmission rate1257 g/m²/24 hTensile strength25.9 lbs./in. (warp), 23.1 lbs./in. (filling)Tearing strength4.2 lbs./in. (lengthwise yarns), 4 lbs./in. (widthwise yarns)Weight48.82 g/m² - White3XL51½"40"Log in for availabilityPallet of 720Pallet of 720 - $8,207.23$8,207.23New Customer Promotional PriceCase of 30$341.97Pallet of 720$8,207.23Specifications:
More
Specifications
Cat. No.414004-382FabricSpunbound polypropyleneClosure typePress studs / snapsCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroom|Industrial|LaboratoryAir permeability840.63 cu. ft./min/sq. ft.Bacterial filtration efficiency55%Particle shedding (Helmke Drum)Level IIWater resistance21.7 GWater vapor transmission rate1257 g/m²/24 hTensile strength25.9 lbs./in. (warp), 23.1 lbs./in. (filling)Tearing strength4.2 lbs./in. (lengthwise yarns), 4 lbs./in. (widthwise yarns)Weight48.82 g/m² - WhiteXXL47½"37½"Log in for availabilityPallet of 720Pallet of 720 - $8,020.03$8,020.03New Customer Promotional PriceCase of 30$334.17Pallet of 720$8,020.03Specifications:
More
Specifications
Cat. No.414004-400FabricSpunbound polypropyleneClosure typePress studs / snapsCleanroom class10000/ISO 7PocketsWithout pocketsGenderUnisexSleeve or leg lengthLongGarment applicationCleanroom|Industrial|LaboratoryAir permeability840.63 cu. ft./min/sq. ft.Bacterial filtration efficiency55%Particle shedding (Helmke Drum)Level IIWater resistance21.7 GWater vapor transmission rate1257 g/m²/24 hTensile strength25.9 lbs./in. (warp), 23.1 lbs./in. (filling)Tearing strength4.2 lbs./in. (lengthwise yarns), 4 lbs./in. (widthwise yarns)Weight48.82 g/m²
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



