Simian NGF (Nerve Growth Factor) ELISA Kit
77525-072
:
- Assay Type:Sandwich
- Host:Monkey
- Target protein:NGF (Nerve Growth Factor)
- Size:96 wells
- Sample type:Serum, plasma, tissue homogenates, cell lysates and other biological fluids.
- Time to Results:3.5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:15.63 - 1000 pg/ml
- Storage temperature:2…8 °C
- Application:Cytokine, neuro science
- Sensitivity:6.2 pg/ml
- Regulatory status:RUO
- Cat. no.:77525-072
- Host: Simian (monkey)