Cattle VEGFA (Vascular Endothelial Growth Factor A) ELISA Kit
Catalog # 77524-724
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Bovine
- Target protein:VEGFA (Vascular Endothelial Growth Factor A)
- Size:96 wells
- Sample type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:15.63 - 1000 pg/ml
- Storage temperature:2…8 °C
- Application:Cytokine, apoptosis, tumor immunity
- Sensitivity:5.9 pg/ml
- Regulatory status:RUO
- Cat. no.:77524-724