Rat RIPK1 (Receptor Interacting Serine Threonine Kinase 1) ELISA Kit
Catalog # 77523-632
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Rat
- Target protein:RIPK1 (Receptor Interacting Serine Threonine Kinase 1)
- Size:96 wells
- Sample type:Tissue homogenates, cell lysates and other biological fluids
- Time to Results:3.5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:0.16 - 10 ng/ml
- Storage temperature:2…8 °C
- Application:Enzyme and kinase
- Sensitivity:0.056 ng/ml
- Regulatory status:RUO
- Cat. no.:77523-632