OFX-100 Lab Coats, Tech Wear
Supplier: Tech Wear
Certificates
About this item
Knee-length lab coats are made of OFX-100 static control fabric and maintain sleeve-to-sleeve continuity for a minimum of 100 washes or two years, whichever comes first
They feature a lapel collar and three pockets. Lab coats are conductive and meet ANSI/ESD STM2.1-1997 garment requirements.
Ordering information: Lab coats are also available grounded and in white and teal; contact your Avantor sales representative for information.
Specifications
- Catalog No:
- 10831-928
- 10831-934
- 10831-922
- 10831-924
- 10831-926
- 10831-920
- 10831-918
- 10831-932
- 10831-930
- Model:
- OFX-100
- OFX-100
- OFX-100
- OFX-100
- OFX-100
- OFX-100
- OFX-100
- OFX-100
- OFX-100
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Fabric:
- OFX-100 static control fabric
- OFX-100 static control fabric
- OFX-100 static control fabric
- OFX-100 static control fabric
- OFX-100 static control fabric
- OFX-100 static control fabric
- OFX-100 static control fabric
- OFX-100 static control fabric
- OFX-100 static control fabric