PYREX® Reducing Adapters with [ST] Joints, Corning
Supplier: Corning
PYREX® Reducing Adapters with [ST] Joints, Corning
Supplier: Corning
Certificates
About this item
PYREX® glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Expansion adapter from small cone to large socket.
3 Options Available Below
Product Details & Documents
PYREX® glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Expansion adapter from small cone to large socket.
Specifications for Product Family
Specifications
- Catalog No:
- 89091-642
- 89091-644
- 62900-088
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
Specifications
Product Family Options
Product Information
- LengthBottom inner [ST] joint sizeTop outer st joint sizeAvailabilityPrice
- Supplier Number: 8800-2419Specifications:Product available on GSA Advantage®
Low Manufacturing Impact More
Specifications
Cat. No.89091-642Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - MonterreyProduct available on GSA Advantage®Low Manufacturing Impact - Supplier Number: 8800-2924Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.89091-644Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - MonterreyLow Manufacturing Impact - Supplier Number: 8800-3424Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.62900-088Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - MonterreyLow Manufacturing Impact