KLEENGUARD® A20 Breathable Particle Protection Lab Coats, Kimberly-Clark Professional®
Supplier: Microflex
Certificates
About this item
Low-linting, MICROFORCE™ Barrier SMS fabric lab coats provide protection with comfort and breathability
Lab coats meet FDA 21 CFR 177.1520 requirements for food contact use. Do not use near open flame. Melting will occur at temperatures higher than 150°C (302°F).
Tailored design with stitched seams, large pockets, and standard collar. Durable, triple-layer construction includes a breathable microfilament matrix layered between tear-resistant outer layers. Snap front.
Specifications
- Catalog No:
- CA10837-080
- CA89066-694
- CA10837-036
- CA10837-069
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Garment application:
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection