PEG (Polyethylene Glycol) ELISA Kit
77525-766
:
- Assay Type:Competitive
- Target protein:PEG (Polyethylene Glycol)
- Size:96 wells
- Sample type:Biological agents
- Time to Results:2 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:15.63 - 1000 ng/ml
- Storage temperature:2…8 °C
- Application:Gastrointestinal science
- Sensitivity:4.72 ng/ml
- Regulatory status:RUO
- Cat. no.:77525-766
- Host: General