Rabbit RLN (Relaxin) ELISA Kit
77523-666
:
- Assay Type:Sandwich
- Host:Rabbit
- Target protein:RLN (Relaxin)
- Size:96 wells
- Sample type:Serum, plasma and other biological fluids
- Time to Results:3.5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:31.25 - 2000 pg/ml
- Storage temperature:2…8 °C
- Application:Endocrinology, Reproductive science, Hormone metabolism
- Sensitivity:9.26 pg/ml
- Regulatory status:RUO
- Cat. no.:77523-666