Human SPTLC3 (Serine Palmitoyltransferase, Long Chain Base Subunit 3) ELISA Kit
Catalog # 77517-834
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Human
- Target protein:SPTLC3 (Serine Palmitoyltransferase, Long Chain Base Subunit 3)
- Size:96 wells
- Sample type:Tissue homogenates, cell lysates and other biological fluids
- Time to Results:3.5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:0.16 - 10 ng/ml
- Storage temperature:2…8 °C
- Application:Enzyme and Kinase
- Sensitivity:0.056 ng/ml
- Regulatory status:RUO
- Cat. no.:77517-834