Rabbit IL8Ra (Interleukin 8 Receptor Alpha) ELISA Kit
Catalog # 77515-266
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Rabbit
- Target protein:IL8Ra (Interleukin 8 Receptor Alpha)
- Size:96 wells
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:0.16 - 10 ng/ml
- Storage temperature:2…8 °C
- Application:Signal transduction, CD and Adhesion molecule, Infection immunity
- Sensitivity:0.055 ng/ml
- Regulatory status:RUO
- Cat. no.:77515-266