Human PRSS1 (Protease, Serine 1) ELISA Kit
77513-490
:
- Assay Type:Sandwich
- Host:Human
- Target protein:PRSS1 (Protease, Serine 1)
- Size:96 wells
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:15.63 - 1000 pg/ml
- Storage temperature:2…8 °C
- Application:Enzyme and Kinase, Infection immunity
- Sensitivity:6.9 pg/ml
- Regulatory status:RUO
- Cat. no.:77513-490