Pig MTOR (Serine/threonine-protein kinase mTOR) ELISA Kit
Catalog # 77526-304
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Pig
- Target protein:MTOR (Serine/threonine-protein kinase mTOR)
- Size:96 wells
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:0.32 - 20 ng/ml
- Storage temperature:2…8 °C
- Application:Cell proliferation
- Sensitivity:0.125 ng/ml
- Regulatory status:RUO
- Cat. no.:77526-304