Human SRSF2 (Serine/Arginine Rich Splicing Factor 2) ELISA Kit
Catalog # 77516-032
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Human
- Target protein:SRSF2 (Serine/Arginine Rich Splicing Factor 2)
- Size:96 wells
- Sample type:Tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:0.16 - 10 ng/ml
- Storage temperature:2…8 °C
- Application:Signal transduction
- Sensitivity:0.054 ng/ml
- Regulatory status:RUO
- Cat. no.:77516-032