Human SPINK1 (Serine Peptidase Inhibitor Kazal Type 1) ELISA Kit
Catalog # 77515-816
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Human
- Target protein:SPINK1 (Serine Peptidase Inhibitor Kazal Type 1)
- Size:96 wells
- Sample type:Serum, plasma and other biological fluids
- Time to Results:3.5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:1.57 - 100 ng/ml
- Storage temperature:2…8 °C
- Application:Tumor immunity
- Sensitivity:0.63 ng/ml
- Regulatory status:RUO
- Cat. no.:77515-816