Rat PRSS1 (Protease, Serine 1) ELISA Kit
Catalog # 77514-100
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Rat
- Target protein:PRSS1 (Protease, Serine 1)
- Size:96 wells
- Sample type:Serum, plasma and other biological fluids
- Time to Results:3.5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:0.63 - 40 ng/ml
- Storage temperature:2…8 °C
- Application:Enzyme and Kinase, Infection immunity
- Sensitivity:0.245 ng/ml
- Regulatory status:RUO
- Cat. no.:77514-100