Mouse IL12A (Interleukin 12A) ELISA Kit
Catalog # 77513-956
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Mouse
- Target protein:IL12A (Interleukin 12A)
- Size:96 wells
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:15.63 - 1000 pg/ml
- Storage temperature:2…8 °C
- Application:Cytokine, Infection immunity
- Sensitivity:5.8 pg/ml
- Regulatory status:RUO
- Cat. no.:77513-956