Mouse SRSF3 (Serine/Arginine Rich Splicing Factor 3) ELISA Kit
Catalog # 77510-788
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Mouse
- Target protein:SRSF3 (Serine/Arginine Rich Splicing Factor 3)
- Size:96 wells
- Sample type:Serum, plasma and other biological fluids
- Time to Results:3.5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:0.16 - 10 ng/ml
- Storage temperature:2…8 °C
- Application:Signal transduction
- Sensitivity:0.071 ng/ml
- Regulatory status:RUO
- Cat. no.:77510-788