Human SPINT2 (Serine Peptidase Inhibitor Kunitz Type 2) ELISA Kit
Catalog # 77516-702
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Human
- Target protein:SPINT2 (Serine Peptidase Inhibitor Kunitz Type 2)
- Size:96 wells
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:0.32 - 20 ng/ml
- Storage temperature:2…8 °C
- Application:Immune molecule
- Sensitivity:0.134 ng/ml
- Regulatory status:RUO
- Cat. no.:77516-702