Zebrafish 5-HT (5-Hydroxytryptamine) ELISA Kit
Catalog # 77527-944
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Competitive
- Host:Zebrafish
- Target protein:5-HT (5-Hydroxytryptamine)
- Size:96 wells
- Sample type:Serum, plasma, tissue homogenates
- Time to Results:2 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:14.07 - 900 ng/ml
- Storage temperature:2…8 °C
- Application:Signal transduction, infection immunity, neuro science, gastroenterology
- Sensitivity:3.97 ng/ml
- Regulatory status:RUO
- Cat. no.:77527-944