Human METRNL (Meteorin Like Protein) ELISA Kit
77524-164
:
- Assay Type:Sandwich
- Host:Human
- Target protein:METRNL (Meteorin Like Protein)
- Size:96 wells
- Sample type:Serum, plasma, tissue homogenates, cell lysates and other biological fluids
- Time to Results:3.5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:1.57 - 100 ng/ml
- Storage temperature:2…8 °C
- Application:Neuro science
- Sensitivity:0.64 ng/ml
- Regulatory status:RUO
- Cat. no.:77524-164