Goat FGF1 (Fibroblast Growth Factor 1, Acidic) ELISA Kit
Catalog # 77521-260
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Goat
- Target protein:FGF1 (Fibroblast Growth Factor 1, Acidic)
- Size:96 wells
- Sample type:Serum, plasma and other biological fluids
- Time to Results:3.5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:78.13 - 5000 pg/ml
- Storage temperature:2…8 °C
- Application:Cytokine, tumor immunity, infection immunity
- Sensitivity:31 pg/ml
- Regulatory status:RUO
- Cat. no.:77521-260