Human IL18BP (Interleukin 18 Binding Protein) ELISA Kit
Catalog # 77518-164
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Human
- Target protein:IL18BP (Interleukin 18 Binding Protein)
- Size:96 wells
- Sample type:Serum, plasma and other biological fluids
- Time to Results:3.5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:1.57 - 100 ng/ml
- Storage temperature:2…8 °C
- Application:Cytokine
- Sensitivity:0.51 ng/ml
- Regulatory status:RUO
- Cat. no.:77518-164