Human LEPR (Leptin Receptor) ELISA Kit
Catalog # 77513-988
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Human
- Target protein:LEPR (Leptin Receptor)
- Size:96 wells
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:31.25 - 2000 pg/ml
- Storage temperature:2…8 °C
- Application:Signal transduction, CD and adhesion molecule, metabolic pathway, endocrinology
- Sensitivity:14.1 pg/ml
- Regulatory status:RUO
- Cat. no.:77513-988