Human TSP-1 (Thrombospondin-1) ELISA Kit
Catalog # 77512-812
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Human
- Target protein:TSP-1 (Thrombospondin-1)
- Size:96 wells
- Sample type:plasma
- Time to Results:3.5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:125 - 8000 pg/ml
- Storage temperature:2…8 °C
- Application:Infection immunity, hematology
- Sensitivity:53 pg/ml
- Regulatory status:RUO
- Cat. no.:77512-812