Human IFABP/FABP2 (Intestinal Fatty Acid Binding Protein) ELISA Kit
77511-972
:
- Assay Type:Sandwich
- Host:Human
- Target protein:IFABP/FABP2 (Intestinal Fatty Acid Binding Protein)
- Size:96 wells
- Sample type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:0.16 - 10 ng/ml
- Storage temperature:2…8 °C
- Application:Metabolic pathway, tumor immunity, endocrinology, cardiovascular biology
- Sensitivity:0.061 ng/ml
- Regulatory status:RUO
- Cat. no.:77511-972