Mouse IL-27 (Interleukin 27) ELISA Kit
Catalog # 77528-314
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Mouse
- Target protein:IL-27 (Interleukin 27)
- Size:96 wells
- Sample type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:31.25 - 2000 pg/ml
- Storage temperature:2…8 °C
- Application:Immunology, cancer
- Sensitivity:18.75 pg/ml
- Regulatory status:RUO
- Cat. no.:77528-314