Rat TAOK2 (Serine/threonine-protein kinase TAO2) ELISA Kit
77526-250
:
- Assay Type:Sandwich
- Host:Rat
- Target protein:TAOK2 (Serine/threonine-protein kinase TAO2)
- Size:96 wells
- Sample type:Serum, plasma and other biological fluids
- Time to Results:3.5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:0.16 - 10 ng/ml
- Storage temperature:2…8 °C
- Application:Functional studies
- Sensitivity:0.058 ng/ml
- Regulatory status:RUO
- Cat. no.:77526-250