Order Entry
Canada
ContactUsLinkComponent
 
Rat PRKAR2b (Protein Kinase, cAMP Dependent Regulatory Type II Beta) ELISA Kit
Catalog # 77525-554
Supplier:  AFG Bioscience
CAS Number:  
undefined
Rat PRKAR2b (Protein Kinase, cAMP Dependent Regulatory Type II Beta) ELISA Kit
Catalog # 77525-554
Supplier:  AFG Bioscience
Supplier Number:  EK247569
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Host:
    Rat
  • Target protein:
    PRKAR2b (Protein Kinase, cAMP Dependent Regulatory Type II Beta)
  • Size:
    96 wells
  • Sample type:
    Tissue homogenates, cell lysates and other biological fluids
  • Time to Results:
    3.5 h
  • Shelf life:
    6 months (4 °C); 12 months (–20 °C)
  • Detection range:
    0.16 - 10 ng/ml
  • Storage temperature:
    2…8 °C
  • Application:
    Enzyme and kinase
  • Sensitivity:
    0.054 ng/ml
  • Regulatory status:
    RUO
  • Cat. no.:
    77525-554

Specifications

About this item