Rat UCN3 (Urocortin 3) ELISA Kit
Catalog # 77524-270
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Competitive
- Host:Rat
- Target protein:UCN3 (Urocortin 3)
- Size:96 wells
- Sample type:Serum, plasma and other biological fluids
- Time to Results:2 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:15.63 - 1000 pg/ml
- Storage temperature:2…8 °C
- Application:Endocrinology
- Sensitivity:4.75 pg/ml
- Regulatory status:RUO
- Cat. no.:77524-270