Rat FUCa1 (Fucosidase Alpha L1, Tissue) ELISA Kit
77519-122
:
- Assay Type:Sandwich
- Host:Rat
- Target protein:FUCa1 (Fucosidase Alpha L1, Tissue)
- Size:96 wells
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:0.32 - 20 ng/ml
- Storage temperature:2…8 °C
- Application:Enzyme and kinase, tumor immunity, hepatology
- Sensitivity:0.115 ng/ml
- Regulatory status:RUO
- Cat. no.:77519-122