Human NRG1 (Neuregulin 1) ELISA Kit
77513-890
:
- Assay Type:Sandwich
- Host:Human
- Target protein:NRG1 (Neuregulin 1)
- Size:96 wells
- Sample type:Tissue homogenates, cell lysates, cell culture supernatants and other biological fluids
- Time to Results:3.5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:0.16 - 10 ng/ml
- Storage temperature:2…8 °C
- Application:Signal transduction, neuro science
- Sensitivity:0.055 ng/ml
- Regulatory status:RUO
- Cat. no.:77513-890