Human PRSS2 (Protease, Serine 2) ELISA Kit
Catalog # 77513-554
Supplier: AFG Bioscience
CAS Number:
Specifications
- Assay Type:Sandwich
- Host:Human
- Target protein:PRSS2 (Protease, Serine 2)
- Size:96 wells
- Sample type:Serum, plasma and other biological fluids
- Time to Results:3.5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:0.79 - 50 ng/ml
- Storage temperature:2…8 °C
- Application:Enzyme and kinase
- Sensitivity:0.28 ng/ml
- Regulatory status:RUO
- Cat. no.:77513-554