Rat Vaspin (Visceral Adipose Tissue Derived Serine Protease Inhibitor) ELISA Kit
77513-482
:
- Assay Type:Sandwich
- Host:Rat
- Target protein:Vaspin (Visceral Adipose Tissue Derived Serine Protease Inhibitor)
- Size:96 wells
- Sample type:Serum, plasma and other biological fluids
- Time to Results:3.5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:93.75 - 6000 pg/ml
- Storage temperature:2…8 °C
- Application:Metabolic pathway, endocrinology
- Sensitivity:35.19 pg/ml
- Regulatory status:RUO
- Cat. no.:77513-482