AccuForge® Single Edge Blades, 3 Facet
Catalog # 77322-480
Supplier: Accutec Blades
Specifications
- Coating:Coated
- Description:Oiled
- Material:Stainless steel
- Thickness:0.009"
- Packed:100 blades/carton, 50 cartons/case
- Cat. no.:77322-480
- Blade Edge:3 Facet
- Backing Material:Aluminum
Specifications
About this item
AccuForge® Single Edge blades feature precision ground edges for sharpness and durability. Blades are wrapped in protective paper.
- 3-Facet edge for increased sharpness
- High-quality stainless steel blade
- Blades are individually wrapped for safety
- Reclosable clamshell for safe and convenient blade storage
- Each clamshell contains 100 blades
- Made in USA
MicroCoat® blade enrichment technology adds a layer of protection for each blade, helping maintain a sharp and effective edge over multiple uses.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.