AccuForge® Double Edge Blades, 400 Blade Dispenser
Catalog # 77322-454
Supplier: Accutec Blades
Specifications
- Coating:Uncoated
- Description:Degreased
- Material:Stainless steel
- Thickness:0.004"
- Packed:400 blades/dispenser, 10 dispensers/case
- Cat. no.:77322-454
- Blade Edge:3 Facet
Specifications
About this item
AccuForge® Double Edge blades feature a 3-Facet edge for exacting, clean cuts.
- 400 blade dispenser for high volume use
- Easy access safety case
- Clear cap for visibility
- 3-Facet blade edge for increased sharpness
- Degreased blade for reduced oil and pariculates
- Made in USA
Paper-free dispenser is great for cleanroom use.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.