PYREX® 75° Connecting/Distilling Adapter with [ST] Joints, Corning
Catalog # 89091-566
Supplier: Corning
Product available on GSA Advantage®
Specifications
- ST Joint:24/40
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Cat. no.:89091-566
- Supplier No.:8930-24
Specifications
About this item
PYREX®, borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
These connecting distilling tube adapters have full-length inner 24/40 standard taper joints of same size. The connecting arm is approximately at a 75° angle from vertical and is 200 mm long.