You searched for: Desiccators
Desiccators create and maintain dry environments to ensure sample stability. Using absorbing beads, and other desiccants, desiccator cabinets or apparatuses quickly absorb moisture, guaranteeing sensitive materials do not react to surroundings prior to testing. Vacuum compatible models accelerate drying of interior conditions. Desiccators are manufactured of clear materials to enable constant monitoring without exposure and may come in UV blocking amber tint. Reusable desiccators are available in a variety of capacities and may feature attached hygrometers.
VWR® Acrylic Desiccator Cabinets
Supplier: VWR International
Acrylic desiccator cabinets provide low-moisture, dust-free storage.
Expand 3 Items
VWR® Large Desiccator Plate
Supplier: VWR International
Chemical-porcelain plate is glazed on one side and contains eight 30 mm holes.
Expand 1 Items
VWR® Vacuum Desiccator, Bottom and Lid with Socket Valve
Supplier: VWR International
The VWR® Vacuum Desiccator is made from high quality Borosilicate 33 expansion glass
Expand 5 Items
VWR® Reusable Silica Gel Desiccant Cartridges
Supplier: VWR International
Useful anywhere desiccation is needed, these cartridges contain inert, non-corrosive, and non-toxic silica gel.
Expand 1 Items
VWR® Humidity Sponges, Desiccator in a Bag
Supplier: VWR International
Humidity sponges turn any container into a desiccator and are ideal for storing samples, food, pharmaceuticals, electronics, PC boards, optics, diagnostics, museum pieces, chemicals, instruments, and documents.
Expand 2 Items
VWR® Desiccator, Bottom and Lid with Glass Knob
Supplier: VWR International
This VWR® Desiccator is made from high quality Borosilicate 33 expansion glass
Expand 5 Items
VWR® Desiccator Plates, Porcelain
Supplier: VWR International
Chemical-porcelain desiccator plates are glazed on one side, perforated with 30 mm diameter holes, and supported by three small feet.
Expand 5 Items
VWR® Desiccator Plates
Supplier: VWR International
Chemical-porcelain dessicator plates are perforated with numerous small holes and a large center hole.
Expand 5 Items
DRIERITE® Mesh Granules
Supplier: VWR
DRIERITE° will absorb 6 to 14% its weight in water from liquids or gases
Expand 3 Items
Granular dessicant, Sicapent® with indicator, MilliporeSigma
Supplier: MilliporeSigma
Sicapent is a granular dessicant comprising ~75% phoshorus pentoxide on an inert inorganic carrier.
Expand 1 Items
DRIERITE® Absorbent, MilliporeSigma
Supplier: MilliporeSigma
Anhydrous calcium sulfate
Expand 6 Items
Desiccator, Plastic, Ace Glass Incorporated
Supplier: Ace Glass
This plastic desiccator has a transparent polycarbonate top, and a white polypropylene bottom
Expand 6 Items
SP Bel-Art "Space Saver" Vacuum Desiccators, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
This small plastic vacuum desiccator is ideal for degasing liquids quickly and easily in the laboratory or classroom.
Expand 6 Items
Vacuum Desiccator with Collar, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
These Vacuum desiccators has a greased rotating collar valve used to facilitate vacuum desiccation
Expand 3 Items
SP Bel-Art Mini Vacuum Desiccator, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
The small size of this desiccator allows quick vacuum creation for rapid, repeated degassing of multiple samples.
Expand 1 Items
Desiccator Cabinet, Ambered, Ace Glass
Supplier: Ace Glass
This stackable, acrylic desiccator cabinet is ambered to protect contents from harmful U.V
Expand 1 Items
SP Bel-Art Secador® 1.0, 2.0, 3.0, 4.0 Vertical Desiccator Cabinets, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
The attractive design and sturdy construction of the Secador® line of desiccator cabinets assures easy and reliable storage of moisture- and light-sensitive products.
Expand 8 Items
SP Bel-Art Grande Desiccator Cabinets, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Extra-large Scienceware® desiccator cabinets provide a low-humidity, dust-free environment for several large containers or a lot more small ones.
Expand 3 Items
DURAN® Vacuum Desiccator, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Vacuum-tight, made from DURAN® borosilicate glass 3.3. To accelerate drying, a vacuum can be applied via the stopcock. Spare parts such as lids, bases, stopcocks and caps can be interchanged (observe DN).
Expand 2 Items
SP Bel-Art Reusable Silica Gel Desiccant Bags, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Convenient, easy to handle packs filled with moisture absorbing silica gel beads are available in multiple sizes to fit most desiccators.
Expand 3 Items
Instatherm Desiccator, Vacuum Oven, Ace Glass Incorporated
Supplier: Ace Glass
The Instatherm vacuum oven desiccator has a heated bottom and an isulated cover
Expand 2 Items
Drying Apparatus, Abderhalden, Ace Glass
Supplier: Ace Glass
When using this improved form Abderhalden vacuum drying apparatus, the material to be dried is placed in the inner tube
Expand 3 Items
Nalgene® Desiccator Cabinets, Acrylic, Thermo Scientific
Supplier: Thermo Fisher Scientific
For storing anhydrous and hygroscopic biological and chemical compounds, as well as electronic assemblies and any components requiring controlled storage conditions.
Expand 3 Items
Auto-Desiccator Cabinets, Ace Glass
Supplier: Ace Glass
This stackable, plastic Secador™ desiccator cabinet is perfect for cramped locations
Expand 1 Items
SP Bel-Art Secador® Refrigerator-Ready Desiccator, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Unique features like an extra-long body and pull-out shelves make the Scienceware® Secador® Refrigerator Ready Desiccator perfect for use inside refrigerators and -20C freezers letting you keep materials cold when needed, and moisture free.
Expand 1 Items
Desiccator Cabinet, Horizontal, Clear, Ace Glass
Supplier: Ace Glass
This stackable, clear plastic Secador™ desiccator cabinet is perfect for cramped locations
Expand 1 Items
SP Bel-Art Reusable Silica Gel Desiccant Cartridges, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Useful anywhere desiccation is needed, these cartridges contain inert, non-corrosive, and non-toxic silica gel.
Expand 1 Items
Nalgene® Desiccator, 150mm, Thermo Scientific
Supplier: Thermo Fisher Scientific
Desiccator cover is clear, shatterproof polycarbonate with exclusive recessed rim design.
Expand 1 Items
SP Bel-Art Vacuum Chamber and Plate, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
This vacuum chamber is useful for vacuum drying, degassing, and teaching demonstrations.