ContactUsLinkComponent
Synthware Distillation Adapters, Kemtech America
Catalog # 60000-656
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Distillation Adapters, Kemtech America
Catalog # 60000-656
Supplier:  Kemtech America
Supplier Number:  A362410
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • ST Joint:
    24/40
  • Cat. no.:
    60000-656
  • Supplier No.:
    A362410
Specifications
About this item
Distillation adapter accommodates a 76 mm immersion thermometer.
  • Accommodates thermometer
  • Thermometer not included
Features a [ST] 10/18 joint on the vertical side tube for a 76 mm immersion thermometer.
Ordering information: Thermometer is not supplied; contact your Avantor representative for more information.